Skip to Content
Merck
GRACE BIO-LABS SEALSTRIPS™ AND SEALSTRIP APPLICATOR, FOR PROPLATE® MICROARRAY SYSTEMS for ProPlate® microarray systems, pack of 1

Grace Bio-Labs SealStrips and SealStrip applicator, for ProPlate® microarray systems

GRACE BIO-LABS SEALSTRIPS™ AND SEALSTRIP APPLICATOR, FOR PROPLATE® MICROARRAY SYSTEMS for ProPlate® microarray systems, pack of 1
Synonym(s):
GRACE BIO-LABS SEALSTRIPS AND SEALSTRIP APPLICATOR, FOR PROPLATE® MICROARRAY SYSTEMS

Products

Shipping
Color
Manufacturer
Package Size
Sterility