Skip to Content
Merck
Glyceryl Monooleate Pharmaceutical Secondary Standard; Certified Reference Material

Glyceryl monooleate

Glyceryl Monooleate Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Glycerol mono-oleate
Empirical Formula (Hill Notation):
C21H40O4
CAS Number:
Molecular Weight:
356.54

Products

Brand
Formula Weight
Manufacturer
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique