Skip to Content
Merck
Gliquidone British Pharmacopoeia (BP) Reference Standard

Gliquidone

Gliquidone British Pharmacopoeia (BP) Reference Standard
Synonym(s):
2-{2-[4-({[(cyclohexylamino)carbonyl]amino}sulfonyl)phenyl]ethyl}-7-methoxy-4,4-dimethyl-1,3-dioxo-1,2,3,4-tetrahydroisoquinoline
Empirical Formula (Hill Notation):
C27H33N3O6S
Molecular Weight:
527.63

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing