Geranylamine
Synonym(s):
trans-3,7-Dimethyl-2,6-octadien-1-ylamine, trans-3,7-Dimethyl-2,6-octadienyl amine
Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCH2NH2
CAS Number:
Molecular Weight:
153.26
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing