Skip to Content
Merck
Galactinol dihydrate analytical standard

Galactinol dihydrate

Galactinol dihydrate analytical standard
Empirical Formula (Hill Notation):
C12H22O11 · 2H2O
CAS Number:
Molecular Weight:
378.33
EC Number:

Products

Product Category
Brand
Formula Weight
Purity
Format
Analyte Suitability
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing