![Gabapentin solution 1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/137/996/1c96aafa-ce3d-4802-a6ee-8820f1f13bea/640/1c96aafa-ce3d-4802-a6ee-8820f1f13bea.png)
Gabapentin solution
![Gabapentin solution 1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/137/996/1c96aafa-ce3d-4802-a6ee-8820f1f13bea/640/1c96aafa-ce3d-4802-a6ee-8820f1f13bea.png)
Synonym(s):
Gabapentin solution
Empirical Formula (Hill Notation):
C9H17NO2
CAS Number:
Molecular Weight:
171.24
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing