Skip to Content
Merck
Gabapentin impurity B European Pharmacopoeia (EP) Reference Standard

Gabapentin impurity B

Gabapentin impurity B European Pharmacopoeia (EP) Reference Standard
Synonym(s):
(1-Cyanocyclohexyl)acetic acid, 1-Cyanocyclohexaneacetic acid
Empirical Formula (Hill Notation):
C9H13NO2
CAS Number:
Molecular Weight:
167.21

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing