![Flurazepam hydrochloride United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/387/516/972908b8-4d05-4f20-b639-8f755b980e26/640/972908b8-4d05-4f20-b639-8f755b980e26.png)
Flurazepam hydrochloride
![Flurazepam hydrochloride United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/387/516/972908b8-4d05-4f20-b639-8f755b980e26/640/972908b8-4d05-4f20-b639-8f755b980e26.png)
Synonym(s):
Flurazepam dihydrochloride, 7-Chloro-1-(diethylamino)ethyl-5-(2-fluorophenyl)-3H-1,4-benzodiazepin-2(1H)-one dihydrochloride
Empirical Formula (Hill Notation):
C21H23ClFN3O · 2HCl
CAS Number:
Molecular Weight:
460.80
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing