![Fenuron certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/191/052/b4ad1030-35fe-4133-9da8-2f091e2b27cc/640/b4ad1030-35fe-4133-9da8-2f091e2b27cc.png)
Fenuron
![Fenuron certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/191/052/b4ad1030-35fe-4133-9da8-2f091e2b27cc/640/b4ad1030-35fe-4133-9da8-2f091e2b27cc.png)
Synonym(s):
1,1-Dimethyl-3-phenylurea, Fenuron
Empirical Formula (Hill Notation):
C9H12N2O
CAS Number:
Molecular Weight:
164.20
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing