Skip to Content
Merck
Fentanyl Related Compound A United States Pharmacopeia (USP) Reference Standard

Fentanyl Related Compound A

Fentanyl Related Compound A United States Pharmacopeia (USP) Reference Standard
Synonym(s):
(2-Bromoethyl)benzene, 2-Phenylethyl bromide
Linear Formula:
C6H5CH2CH2Br
CAS Number:
Molecular Weight:
185.06

Products

Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing