
Fenpropidin Metabolite CGA 289267

Synonym(s):
2-Methyl-2-(4-(2-methyl-3-piperidin-1-ylpropyl)phenyl)propionic acid
Empirical Formula (Hill Notation):
C19H29NO2
CAS Number:
Molecular Weight:
303.44
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing