Skip to Content
Merck
Farnesol mixture of isomers, analytical standard

Farnesol

Farnesol mixture of isomers, analytical standard
Synonym(s):
3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol
Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCH2CH2C(CH3)=CHCH2OH
CAS Number:
Molecular Weight:
222.37
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique