Etodolac impurity H
Synonym(s):
2-(7-Ethylindol-3-yl)ethanol
Empirical Formula (Hill Notation):
C12H15NO
CAS Number:
Molecular Weight:
189.25
Products
Formula Weight
Manufacturer
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing