
Etiocholan-3α-ol-17-one

Synonym(s):
3α-Hydroxy-5β-androstan-17-one, 5β-Androsterone, Etiocholanone
Empirical Formula (Hill Notation):
C19H30O2
CAS Number:
Molecular Weight:
290.44
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing