Skip to Content
Merck
Ethyl crotonate 99%

Ethyl crotonate

Ethyl crotonate 99%
Linear Formula:
CH3CH=CHCOOC2H5
Molecular Weight:
114.14
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing