Skip to Content
Merck
Ethyl 2-methyl-1,3-dioxolane-2-acetate ≥99%

Ethyl 2-methyl-1,3-dioxolane-2-acetate

Ethyl 2-methyl-1,3-dioxolane-2-acetate ≥99%
Synonym(s):
Fructone, 2-Methyl-1,3-dioxolane-2-acetic acid ethyl ester, Applinal, Ethyl 2-methyldioxolan-2-yl acetate, Jasmaprunat, NSC 6537
Empirical Formula (Hill Notation):
C8H14O4
CAS Number:
Molecular Weight:
174.19

Products

Product Category
Brand
Biological Source
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing