Skip to Content
Merck
Estazolam United States Pharmacopeia (USP) Reference Standard

Estazolam

Estazolam United States Pharmacopeia (USP) Reference Standard
Synonym(s):
8-Chloro-6-phenyl-4H-[1,2,4]triazolo[4,3-a][1,4]benzodiazepine
Linear Formula:
C16H11N4Cl
CAS Number:
Molecular Weight:
294.74

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing