Eprinomectin
Synonym(s):
Eprinomectin, (4′’R)-4″-(Acetylamino)-4″-deoxy-avermectin B1
Empirical Formula (Hill Notation):
C50H75NO14
CAS Number:
Molecular Weight:
914.13
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique