Skip to Content
Merck
Entacapone ≥98% (HPLC)

Entacapone

Entacapone ≥98% (HPLC)
Synonym(s):
2-Cyano-N,N-diethyl-3-(3,4-dihydroxy-5-nitrophenyl)propenamide
Empirical Formula (Hill Notation):
C14H15N3O5
CAS Number:
Molecular Weight:
305.29

Products

Product Category
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade