Elastase from porcine pancreas
Synonym(s):
Elastase from hog pancreas, Pancreatopeptidase E
CAS Number:
EC Number:
Products
Product Category
Brand
Biological Source
Physical Form
Tissue
Chemical Composition
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing