
Ecamsule Related Compound C

Synonym(s):
Terephthalic acid, Benzene-1,4-dicarboxylic acid
Linear Formula:
C6H4-1,4-(CO2H)2
CAS Number:
Molecular Weight:
166.13
Products
Formula Weight
Manufacturer
Melting Point (°C)
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing