Skip to Content
Merck
D-Threitol 99%

D-Threitol

D-Threitol 99%
Synonym(s):
(2R,3R)-1,2,3,4-Butanetetrol
Linear Formula:
HOCH2[CH(OH)]2CH2OH
CAS Number:
Molecular Weight:
122.12

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing