(+)-Terpinen-4-ol
Synonym(s):
(S)-1-Isopropyl-4-methyl-3-cyclohexen-1-ol, (S)-p-Menth-1-en-4-ol
Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Products
Brand
Boiling Point (°C)
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing