
(+)-Taxifolin

Synonym(s):
Taxifolin, (2R,3R)-3,3′,4′,5,7-Pentahydroxyflavanone, (2R,3R)-Dihydroquercetin
Empirical Formula (Hill Notation):
C15H12O7
CAS Number:
Molecular Weight:
304.25
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1