![DMT-2′O-TC-rG(ib) Phosphoramidite configured for PerkinElmer, configured for PolyGen](/deepweb/assets/sigmaaldrich/product/structures/179/935/d6a94439-12e9-45a7-97d3-0eec50f06efa/640/d6a94439-12e9-45a7-97d3-0eec50f06efa.png)
DMT-2′O-TC-rG(ib) Phosphoramidite
![DMT-2′O-TC-rG(ib) Phosphoramidite configured for PerkinElmer, configured for PolyGen](/deepweb/assets/sigmaaldrich/product/structures/179/935/d6a94439-12e9-45a7-97d3-0eec50f06efa/640/d6a94439-12e9-45a7-97d3-0eec50f06efa.png)
Synonym(s):
N2-Isobutyryl-O5′-(4,4′-dimethoxytrityl)-O2′-(1,1-dioxothiomorpholine-4-thiocarbonyl)guanosine O3′-(O-(2-cyanoethyl)-N,N-diisopropylphosphoramidite)
Empirical Formula (Hill Notation):
C49H61N8O11PS2
CAS Number:
Molecular Weight:
1033.16
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Showing 1-3 of 3