![(±)-MDA-D5 solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/180/312/ed423683-8611-4f60-988a-74982d13dad6/640/ed423683-8611-4f60-988a-74982d13dad6.png)
(±)-MDA-D5 solution
![(±)-MDA-D5 solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/180/312/ed423683-8611-4f60-988a-74982d13dad6/640/ed423683-8611-4f60-988a-74982d13dad6.png)
Empirical Formula (Hill Notation):
C10D5H8NO2
CAS Number:
Molecular Weight:
184.25
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2