![Diflorasone diacetate United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/299/243/e7b1a9b2-4685-43a4-ad6a-28d8f78b3543/640/e7b1a9b2-4685-43a4-ad6a-28d8f78b3543.png)
Diflorasone diacetate
![Diflorasone diacetate United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/299/243/e7b1a9b2-4685-43a4-ad6a-28d8f78b3543/640/e7b1a9b2-4685-43a4-ad6a-28d8f78b3543.png)
Synonym(s):
6α,9-Difluoro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione 17,21-diacetate
Empirical Formula (Hill Notation):
C26H32F2O7
CAS Number:
Molecular Weight:
494.52
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing