Diethylpropion hydrochloride solution
Empirical Formula (Hill Notation):
C13H20ClNO
CAS Number:
Molecular Weight:
241.76
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing