
Dichloro(1,10-phenanthroline)palladium(II)

Empirical Formula (Hill Notation):
C12H8Cl2N2Pd
CAS Number:
Molecular Weight:
357.53
Products
Brand
Formula Weight
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing