Skip to Content
Merck
Dicarbonylcyclopentadienyl cobalt(I) technical grade

Dicarbonylcyclopentadienyl cobalt(I)

Dicarbonylcyclopentadienyl cobalt(I) technical grade
Synonym(s):
Cobalt dicarbonyl cyclopentadiene, Cyclopentadienyl cobalt(I) dicarbonyl, Cyclopentadienylcobalt dicarbonyl, Cyclopentadienyldicarbonylcobalt
Linear Formula:
C5H5Co(CO)2
CAS Number:
Molecular Weight:
180.05
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Reagent Type
Core
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing