Dibutylboryl trifluoromethanesulfonate solution
Synonym(s):
Dibutylboron triflate solution
Linear Formula:
CF3SO3B[(CH2)3CH3]2
CAS Number:
Molecular Weight:
274.11
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing