Dexamethasone phosphate
Synonym(s):
(11β,16α)-9-Fluoro-11,17-dihydroxy-16-methyl-21-(phosphonooxy)pregna-1,4-diene-3,20-dione, Dexamethasone phosphate
Empirical Formula (Hill Notation):
C22H30FO8P
Molecular Weight:
472.44
EC Number:
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2