Skip to Content
Merck
Desipramine-D3 hydrochloride solution 1.0 mg/mL in methanol (as free base), certified reference material, Cerilliant®

Desipramine-D3 hydrochloride solution

Desipramine-D3 hydrochloride solution 1.0 mg/mL in methanol (as free base), certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C18D3H19N2 · HCl
CAS Number:
Molecular Weight:
305.86

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique