![Delta6-testosterone certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/981/315/b9f5b969-2c01-4077-ba33-477d0ac3d041/640/b9f5b969-2c01-4077-ba33-477d0ac3d041.jpg)
Delta6-testosterone
![Delta6-testosterone certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/981/315/b9f5b969-2c01-4077-ba33-477d0ac3d041/640/b9f5b969-2c01-4077-ba33-477d0ac3d041.jpg)
Synonym(s):
17β-Hydroxyandrosta-4,6-dien-3-one, 4,6-Androstadiene-17β-ol-3-one
Empirical Formula (Hill Notation):
C19H26O2
CAS Number:
Molecular Weight:
286.41
Products
Brand
Formula Weight
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing