
Decaethylene glycol monododecyl ether

Synonym(s):
C12E10, Polyoxyethylene (10) lauryl ether
Empirical Formula (Hill Notation):
C32H66O11
CAS Number:
Molecular Weight:
626.86
Products
Shipping
Product Category
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing