Skip to Content
Merck
δ-Dodecalactone mixture of isomers, natural, ≥98%, FG

δ-Dodecalactone

δ-Dodecalactone mixture of isomers, natural, ≥98%, FG
Synonym(s):
(±)-5-Dodecanolide, (±)-δ-Heptyl-δ-valerolactone, (±)-6-Heptyltetrahydro-2H-pyran-2-one, 5-Hydroxydodecanoic acid δ-lactone
Empirical Formula (Hill Notation):
C12H22O2
CAS Number:
Molecular Weight:
198.30
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing