DavePhos-Pd-G3
Synonym(s):
Methanesulfonato 2-dicyclohexylphosphino-2-(N,N-dimethylamino)biphenyl(2′-amino-1,1′-biphenyl-2-yl) palladium(II)
Empirical Formula (Hill Notation):
C39H49N2O3PPdS
CAS Number:
Molecular Weight:
763.28
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing