Skip to Content
Merck
vibrant-m

Cyclobenzaprine Related Compound B

vibrant-m
Synonym(s):
N-Desmethylcyclobenzaprine hydrochloride, 3-(5H-Dibenzo[a,d]cyclohepten-5-ylidene)-N-methyl-1-propanamine hydrochloride, N-Methyl-3-(2-tricyclo[9.4.0.03,8]pentadeca-1(15),3,5,7,9,11,13-heptaenylidene)propan-1-amine hydrochloride, Norproheptatriene hydrochloride
Empirical Formula (Hill Notation):
C19H19N · HCl
CAS Number:
Molecular Weight:
297.82

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1