Skip to Content
Merck
vibrant-m

Cyclobenzaprine Related Compound A

vibrant-m
Synonym(s):
5-[3-(Dimethylamino)propyl]-5H-dibenzo[a,d]cyclohepten-5-ol
Empirical Formula (Hill Notation):
C20H23NO
CAS Number:
Molecular Weight:
293.40

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade