Skip to Content
Merck
Clotrimazole British Pharmacopoeia (BP) Reference Standard

Clotrimazole

Clotrimazole British Pharmacopoeia (BP) Reference Standard
Synonym(s):
1-(o-Chloro-α,α-diphenylbenzyl)imidazole, 1-(o-Chlorotrityl)imidazole, 1-[(2-Chlorophenyl)diphenylmethyl]-1H-imidazole
Empirical Formula (Hill Notation):
C22H17ClN2
CAS Number:
Molecular Weight:
344.84
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique