Skip to Content
Merck
cis-3-Hexenyl pyruvate ≥93%

cis-3-Hexenyl pyruvate

cis-3-Hexenyl pyruvate ≥93%
Synonym(s):
3-Hexenyl 2-oxopropanoate
Empirical Formula (Hill Notation):
C9H14O3
CAS Number:
Molecular Weight:
170.21

Products

Product Category
Brand
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Showing 1-1 of 1
Compare
Product Number
Description
Pricing