Cinidon-ethyl
Synonym(s):
cis-Ethyl 2-chloro-3-[2-chloro-5-(1,3,4,5,6,7-hexahydro-1,3-dioxo-2H-isoindol-2-yl)phenyl]-2-propenoate
Empirical Formula (Hill Notation):
C19H17Cl2NO4
CAS Number:
Molecular Weight:
394.25
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing