Chlordiazepoxide Hydrochloride CIV
Synonym(s):
Chlordiazepoxide hydrochloride, 7-Chloro-2-(methylamino)-5-phenyl-3H-1,4-benzodiazepine 4-oxide hydrochloride
Empirical Formula (Hill Notation):
C16H14ClN3O · HCl
CAS Number:
Molecular Weight:
336.22
Products
Brand
Formula Weight
Agency
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing