Skip to Content
Merck
Chlordiazepoxide hydrochloride European Pharmacopoeia (EP) Reference Standard

Chlordiazepoxide hydrochloride

Chlordiazepoxide hydrochloride European Pharmacopoeia (EP) Reference Standard
Synonym(s):
7-Chloro-2-(methylamino)-5-phenyl-3H-1,4-benzodiazepine 4-oxide hydrochloride
Empirical Formula (Hill Notation):
C16H14ClN3O · HCl
CAS Number:
Molecular Weight:
336.22

Products

Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade