Cefdinir
Synonym(s):
syn-7-[2-(2-amino-4-thiazolyl)-2-hydroxyiminoacetamido]-3-vinyl-3-cephem-4-carboxylic acid, BMY-28488, FK-482, [6R-[6α-7beta(Z)]]-7-[[(2-Amino-4-thiazolyl)(hydroxyimino)acetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Empirical Formula (Hill Notation):
C14H13N5O5S2
CAS Number:
Molecular Weight:
395.41
Products
Brand
Formula Weight
Physical Form
pK Value
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing