![Cefadroxil Related Compound B pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/175/507/dbe82632-308a-47df-8d80-18a6f941d084/640/dbe82632-308a-47df-8d80-18a6f941d084.png)
Cefadroxil Related Compound B
![Cefadroxil Related Compound B pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/175/507/dbe82632-308a-47df-8d80-18a6f941d084/640/dbe82632-308a-47df-8d80-18a6f941d084.png)
Synonym(s):
7-Aminodesacetoxycephalosporanic acid, 7-ADCA
Empirical Formula (Hill Notation):
C8H10N2O3S
CAS Number:
Molecular Weight:
214.24
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2