![Carbenicillin sodium European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/155/541/2a9e691c-2555-436d-b761-ecac22aacc2a/640/2a9e691c-2555-436d-b761-ecac22aacc2a.png)
Carbenicillin sodium
![Carbenicillin sodium European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/155/541/2a9e691c-2555-436d-b761-ecac22aacc2a/640/2a9e691c-2555-436d-b761-ecac22aacc2a.png)
Synonym(s):
Carbenicillin disodium salt, α-Carboxybenzylpenicillin disodium salt
Empirical Formula (Hill Notation):
C17H16N2Na2O6S
CAS Number:
Molecular Weight:
422.36
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing