![Buparvaquone VETRANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/331/602/d4b93f79-bae6-437d-88e3-b0f5d7b76e91/640/d4b93f79-bae6-437d-88e3-b0f5d7b76e91.png)
Buparvaquone
![Buparvaquone VETRANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/331/602/d4b93f79-bae6-437d-88e3-b0f5d7b76e91/640/d4b93f79-bae6-437d-88e3-b0f5d7b76e91.png)
Synonym(s):
2-[(4-tert-Butylcyclohexyl)methyl]-3-hydroxy-1,4-naphthoquinone, 2-[[4-(1,1-Dimethylethyl)cyclohexyl]methyl]-3-hydroxy-1,4-naphthalenedione
Empirical Formula (Hill Notation):
C21H26O3
CAS Number:
Molecular Weight:
326.43
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing