
Budesonide Related Compound L

Synonym(s):
16α,17-[Butylidenebis(oxy)]-21-hydroxypregna-1,4-diene-3,11,20-trione, 11-Ketobudesonide
Empirical Formula (Hill Notation):
C25H32O6
CAS Number:
Molecular Weight:
428.52
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2