![Brevetoxin 2 solution 20 μg/g in acetonitrile (nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/280/952/99406456-b999-405b-b492-b4278779cf3e/640/99406456-b999-405b-b492-b4278779cf3e.png)
Brevetoxin 2 solution
![Brevetoxin 2 solution 20 μg/g in acetonitrile (nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/280/952/99406456-b999-405b-b492-b4278779cf3e/640/99406456-b999-405b-b492-b4278779cf3e.png)
Synonym(s):
Brevetoxin B solution
Empirical Formula (Hill Notation):
C50H70O14
CAS Number:
Molecular Weight:
895.08
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1